Learn More
Fisherbrand™ Accumet™ Temperature-Compensated Two-Cell Conductivity Probes
Product Code. 11849043 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Connector Types:
8-pin
Unit Size:
Each
Product Code. Connector Types Unit Size
11849043 8-pin Each
1 options
Product Code. 11849043
  • ONLINE EXCLUSIVE PRICE
    600.00 €
    426.63 € / Pack of 1
    Save 173.37 €
    28% Off
View all promotions
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Fisherbrand™ Accumet™ Temperature-Compensated Two-Cell Conductivity Probes - Mercury - Free, k=10cm-1, 1 to 200 mS/cm, with 8 pin connector
Description
For use on Accumet™ Excel (XL) series benchtop meters
  • Immersion-type cell
  • Use in standard electrode holder: unique compact size is same as typical pH electrode
  • Built-in temperature sensors provide temperature compensated readings
  • Epoxy body
TRUSTED_SUSTAINABILITY
Specifications
Cell Constant 10
Connector Types 8-pin
Sensor Types Conductivity
Temperature Accuracy Temperature Compensated
Type Conductivity Probe
Material Epoxy
Measurement Ranges 1 to 200 mS/cm