missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Autoclavable Biohazard Waste Bags

Product Code. 18336533 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity (English):
13 to 16 gal.
20 to 30 gal.
44 gal.
7 to 10 gal.
8 to 12 gal.
Dimensions (L x W):
23 x 19 in.
30 x 24 in.
35 x 25 in.
38 x 31 in.
48 x 37 in.
Unit Size:
Pack of 100
Pack of 200
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (English) Dimensions (L x W) Unit Size
18336533 20 to 30 gal. 38 x 31 in. Pack of 200
18558056 7 to 10 gal. 23 x 19 in. Pack of 100
18306513 13 to 16 gal. 35 x 25 in. Pack of 100
18366523 8 to 12 gal. 30 x 24 in. Pack of 200
18316533 44 gal. 48 x 37 in. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 18336533
  • ONLINE EXCLUSIVE PRICE
    548.10 €
    196.07 € / Pack of 200
    Save 352.03 €
    64% Off
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Dual tested compliant.

Dual tested compliant

 Autoclaving is a very common way to sterilize infectious waste through the use of high temperature, high pressure steam over a specific time period. Sterilized waste is less expensive to dispose. Dual-tested autoclave bags are ASTM D1922 Elmendorf Tear tested to exceed 480g of tear strength (MD and TD) and ASTM D1709 tested to exceed 165g dart drop strength.

  • Integral steam processing indicator that turns brown when autoclaved
  • Bags comply with US DOT 49CFR Sections 173-197 and specific state regulations
  • Operating temperature tolerance up to 285degreeFdegreeAvailable in various sizes
  • Bag with black print
  • Gauge: 2mil
Note: Not compliant with Delaware, which requires 3 mil; check state regulations.

Specifications

Capacity (English) 20 to 30 gal.
Color Red, Black Print
Dimensions (L x W) 38 x 31 in.
Length (Metric) 96.5 cm
Width (Metric) 78.7 cm
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.