missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Sterile Cell Strainers

Product Code. 11587522 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Mesh Size:
40 μm
70 μm
100 μm
Color:
Blue
White
Yellow
Unit Size:
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mesh Size Color Unit Size
11587522 40 μm Blue Pack of 50
11597522 70 μm White Pack of 50
11517532 100 μm Yellow Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11587522
Spotlight Offers
Use promo code 28036
View all promotions
In Stock
Add to basket
Edge
Add to basket

Sterile and ready to use for straining stem and primary cell samples. Disposable strainers feature universal fit for compatibility with most 50mL conical tubes. Available in three color-coded mesh sizes.

Rapidly isolate primary cells for single-cell suspensions using Fisherbrand™ Cell Strainers. Sterile and ready to use for straining stem and primary cell samples. Disposable strainers feature universal fit for compatibility with most 50mL conical tubes. Available in three color-coded mesh sizes.

Rapidly isolate primary cells
  • Fast, simple alternative to gauze filtration
  • Reliably dissociates cells from clumps or primary tissues
  • Consistently obtain a uniform single-cell suspension
  • Ideal for preparation of:
    • Single-cell suspensions of stem and primary (blood, tissue, cancer) cells
    • Specimens for primary cell cultures and immunogens
    • Freezing stocks
    • Filtering agglutinative proteins produced in inactivation serum
Reliable and easy to use
  • Strong nylon mesh with evenly spaced pores for consistent results
  • Sterilized by gamma irradiation and non-cytotoxic to maintain sample integrity
  • Includes an extended lip for easier handling with forceps
  • Color-coded polypropylene frame with tab enables easy handling and identification
  • Individually packed, ready to use and disposable

Specifications

Mesh Material Nylon
Disposable Single-use
Color Blue
Material Polypropylene
Product Type Cell Strainer
Sterility Sterile
For Use With (Equipment) Conical Tubes, 50 mL
Mesh Size 40 μm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.