missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Color Coders for Externally and Internally Threaded Cryogenic Storage Vials

Product Code. 11977974 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
For externally threaded vials
For internally threaded vials
Color:
Blue
Green
Red
White
Yellow
Unit Size:
Pack of 500
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Color unitSize
11977974 For internally threaded vials White Pack of 500
11927974 For externally threaded vials White Pack of 500
11937974 For externally threaded vials Yellow Pack of 500
11947974 For externally threaded vials Green Pack of 500
11957974 For externally threaded vials Blue Pack of 500
11967974 For externally threaded vials Red Pack of 500
11987974 For internally threaded vials Yellow Pack of 500
11997974 For internally threaded vials Green Pack of 500
11907984 For internally threaded vials Blue Pack of 500
11917984 For internally threaded vials Red Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 11977974
Add to basket
Add to basket

For sample identification

Fit into the caps of Fisherbrand Externally Threaded Cryogenic Storage Vials (10-500-25, -26, and -27) or Internally Threaded Cryogenic Storage Vials (12-567-500, -501, -502).

  • For color-coded sample identification
  • Polystyrene
  • Nonsterile

TRUSTED_SUSTAINABILITY

Specifications

Color White
Material Polystyrene
Sterility Non-sterile
For Use With (Equipment) For internally threaded vials
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.