missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Lightweight Cotton Lisle Inspection Gloves

Product Code. 30280310 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Size:
Jumbo
Medium
Unit Size:
Pack of 12
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
30280310 Jumbo Pack of 12
12865393 Medium Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30280310
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ideal for general inspection work, or for use as liners underneath unlined liquid proof gloves.

  • 100% cotton lisle gloves
  • Two-piece construction
  • Wear alone for inspection purposes
  • Can be used as liners under liquid proof gloves
  • Ambidextrous

Specifications

Construction Cut and Sewn
Coating Material Uncoated
Color White
Disposable Single-use
Hand Ambidextrous
Latex Free Yes
Material Cotton
Product Type Inspection Gloves
Size Jumbo
Sterility Non-sterile
For Use With (Application) Inspection, Electronics Assembly
Gender Men's
Length (English) 9 in.
Length (Metric) 36 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.