missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Dispenser Tips for Handystep™ S Repeating Pipetter

Product Code. 17833172 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Volume (Metric):
0.5 mL
1 mL
2.5 mL
Sterility:
Sterile
Unit Size:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Sterility Unit Size
17833172 2.5 mL Sterile Pack of 100
17433573 0.5 mL Sterile Pack of 100
17823172 1 mL Sterile Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17833172
Save Up to 60% on Liquid Handling
Use promo code 28028
View all promotions
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Manufactured from high-quality virgin plastics for use with most standard repeating pipettes in a wide variety of applications. The tips are made without chemical additives to prevent reagent contamination.

Manufactured from high-quality virgin plastics for use with all standard repeating pipettes in a wide variety of applications. The tips are made without chemical additives to prevent reagent contamination.

25 mL and 50 mL tips require the use of an adapter. One reusable adapter supplied in each package of 25 mL and 50 mL tips.

  • Precision seal between the piston and cylinder ensure smooth operation, increased accuracy and precision
  • Extended piston design minimizes dead volume and maximizes fluid utilization
  • Made entirely from high-quality virgin, high density polyethylene and polypropylene (piston of 0.1 mL tip is liquid crystal polymer-LCP)

Available sterile or nonsterileFits these models but not limited to:

  • BRAND™ HandyStep™, HandyStep™ S, HandyStep™ electronic, HandyStep™ touch, HandyStep™ touch S
  • All previous Fisherbrand Repeating Pipettes
  • Rainin AutoRep™ M, AutoRep™ S, AutoRep™ E
  • Eppendorf Multipette™ 4780
  • Gilson Repetman™
  • DLAB StepMate
  • Scilogex Repeater

Specifications

Volume (Metric) 2.5 mL
Volume (Metric) Dispensing 500 to 2500μL
Filtered Non-filtered
Sterility Sterile
Color Translucent
Autoclavable Not Autoclavable
Precision ≤0.8 to 0.2%
Accuracy ±≤3.5 to 0.7%
Clean Claims Free from endotoxins, DNA, RNase, DNase, and ATP
Sterilization Method Beta
Format Individually Wrapped
Tip Style Positive Displacement
Quantity 100/Pk.
Product Type Syringe Tips
Disposable Single-use
For Use With (Equipment) Most repeating pipette models (see Description), Fisherbrand HandyStep S
Graduated Yes
Material HDPE, Polypropylene
Graduations Printed
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.