missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Flat and Domed 8-Cap Strips, Flat Cap Strips

Product Code. 11859251 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Format:
Domed Cap Strips
Flat Cap Strips
Unit Size:
Pack of 250
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format Unit Size
11859251 Domed Cap Strips Pack of 250
12184102 Flat Cap Strips Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11859251
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Suitable for use with 96-well PCR plates and PCR tube strips

  • Eight caps per strip
  • Cleanroom produced
  • Virgin polypropylene
  • Flat caps offer large, labeled end tabs for easy removal and strip orientation

Specifications

Material Polypropylene
Color Natural
Format Domed Cap Strips
For Use With (Equipment) 96-well PCR Plates or PCR Tube Strips
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.