Learn More
Fisherbrand™ General Filtration Membrane Filters
Useful for most filter applications
Brand: Fisherbrand™
Additional Details : Weight : 0.30000kg
Description
- Plain, white membrane filters
- Pure, biologically inert mixture of cellulose acetate and cellulose nitrate (MCE)
- Compatible with dilute acids and bases, aliphatic and aromatic hydrocarbons, and nonpolar liquids
- Filters wet readily with water and other fluids
- Contain no fibers or particles to contaminate filtrate
- Autoclavable
Applications:
For all analytical, ultracleaning, and sterilizing applications at temperatures below 75°C (167°F)—except those involving esters, ether-alcohols, ketones, nitroparaffins or strong acids and bases
Specifications
Autoclavable | |
0.45 μm | |
25 mm |
MCE | |
For all analytical, ultracleaning, and sterilizing applications at temperatures below 75°C (167°F) – except those involving esters, ether-alcohols, ketones, nitroparaffins or strong acids and bases. |