missing translation for 'onlineSavingsMsg'
Learn More

Humidity Sponge™, Fisherbrand™

Product Code. 11734366 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Chemical Name or Material:
Fisherbrand™ Humidity Sponge™ Dessicant
Unit Size:
Pack of 40
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Chemical Name or Material Unit Size
11734366 Fisherbrand™ Humidity Sponge™ Dessicant Pack of 40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11734366
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Humidity Sponge Dessicant; Fisherbrand; Reusable Type

  • Non-toxic desiccant
  • Reusable
  • Material is made of DuPont's Tyvek™ breathable, high-density polyethylene
  • Perfect for drugs, food, electronics, optics and equipment
  • Bags are guaranteed to retain 16% moisture by weight, typical results are 37% moisture absorption
  • Desiccant meets US FDA requirements
  • Certified ISO Class 5 (Class M3.5) cleanroom compatible

Specifications

Chemical Name or Material Fisherbrand™ Humidity Sponge™ Dessicant
CAS 1302-78-9
Material HDPE
Quantity 40 Pk.
Grade Certified
Weight 11 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.