missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Humidity Sponge™ Dessicant Window Type, Fisherbrand™
Shop All Cole-Parmer Products
Click to view available options
Chemical Name or Material:
Fisherbrand™ Humidity Sponge™ Dessicant
Unit Size:
Pack of 40
Description
Meet all drying requirements with these humidity sponges. Fisherbrand™ Dessicant Window Type Humidity Sponges™ are non-toxic and Class M3.5 cleanroom-compatible.
- Bags result in 37% moisture absorption
- Indicators show when moisture capacity has been reached
- One bag will retain 16% moisture in a container with a volume of two gallons (476 cu. in.)
- Bag size: 7.6 x 7.6 x 0.6 cm (3 x 3 x 1/4 in.)
Specifications
Specifications
| Chemical Name or Material | Fisherbrand™ Humidity Sponge™ Dessicant |
| CAS | 1302-78-9 |
| Material | Sodium Calcium Aluminosilicate Hydrate |
| Quantity | 40 Pk. |
| Grade | Certified |
| Weight | Each bag weighs 10 g (0.35 oz.) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction