Learn More
Fisherbrand™ Lens Paper
Product Code. p-95151 Shop All Fisherbrand Products
Change view
Click to view available options
Dimensions (L x W):
4 x 6 in.
6 x 8 in. sheets
Quantity:
100 Sheets/Book, 12 Books/Pk.
50 Sheets/Book, 12 Books/Pk.
Unit Size:
Pack of 12
Pack of 600
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Quantity unitSize
15392267 6 x 8 in. sheets 100 Sheets/Book, 12 Books/Pk. Pack of 12
10216202 4 x 6 in. 50 Sheets/Book, 12 Books/Pk. Pack of 600
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15392267
Only null left
Add to basket
Add to basket
For cleaning glass lenses
Booklet
  • Never scratches glass lenses, objectives, or other delicate surfaces
  • Doesn't become harsh; doesn't readily collect dust or grease
  • Virtually lint-free sheets are glued and bound in a booklet for easy removal
Dispenser Box
  • Same paper as above, but packaged in a convenient dispenser box for fast, single-sheet removal individual sheets measure 5L × 7 in.W (13 × 18cm)
Specifications
Product Type Lens Cleaning Paper
Dimensions (L x W) 6 x 8 in. sheets
For Use With (Application) For cleaning Glass lenses
Quantity 100 Sheets/Book, 12 Books/Pk.