missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Black Phenolic PolyCone Cap

Product Code. 15502361 Shop All Fisherbrand Products
Change view
Click to view available options
Neck Finish:
20-400
28-400
Unit Size:
Pack of 576
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Neck Finish unitSize
15502361 28-400 Pack of 576
11528172 20-400 Pack of 576
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15502361

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Forms an exceptionally tight seal and offers a good chemical barrier. Fisherbrand™ Black Phenolic PolyCone Caps are crack resistant.

PolyCone Lined Phenolic Caps form an exceptionally tight seal and offer a good chemical barrier. This liner molds itself around the sealing areas of the finish as the cap is screwed on, virtually eliminating leakage, evaporation, contamination, binding and backing off. Great for everything from liquids, to soaps, to macro-invertebrates.

  • Liner molds itself around sealing areas of the finish as the cap is screwed on, virtually eliminating leakage, evaporation, contamination, binding and backing off
  • Great for everything from liquids, to soaps, to macro-invertebrates

Specifications

Color Black
Description Cap
Material Phenolic Resin
No. per Case 48 Cs.
Packaging Packed in 48 bags of 12
Quantity 576EA/Cs.
Liner Material PolyCone
Neck Finish 28-400
Certifications/Compliance ISO 9001
Shape Round
Product Type Screw Cap
For Use With (Equipment) Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.