missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Black Phenolic PolyCone Cap

Product Code. p-2664189 Shop All Fisherbrand Products
Click to view available options
Neck Finish:
20-400
28-400
Unit Size:
Pack of 576
This item is not returnable. View return policy

Product Code. 11528172

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Forms an exceptionally tight seal and offers a good chemical barrier. Fisherbrand™ Black Phenolic PolyCone Caps are crack resistant.

PolyCone Lined Phenolic Caps form an exceptionally tight seal and offer a good chemical barrier. This liner molds itself around the sealing areas of the finish as the cap is screwed on, virtually eliminating leakage, evaporation, contamination, binding and backing off. Great for everything from liquids, to soaps, to macro-invertebrates.

  • Liner molds itself around sealing areas of the finish as the cap is screwed on, virtually eliminating leakage, evaporation, contamination, binding and backing off
  • Great for everything from liquids, to soaps, to macro-invertebrates

Specifications

Color Black
Material Phenolic Resin
No. per Case 576 Cs.
Packaging Packed in 48 bags of 12
Quantity 576EA/Cs.
Liner Material PolyCone
Neck Finish 20-400
Certifications/Compliance ISO 9001
Shape Round
Product Type Screw Cap
For Use With (Equipment) Bottle
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.