missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Sterile PETG Media Bottles
GREENER_CHOICE

Product Code. 12084118 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1000 mL
Closure Type:
Tamper-evident Screw Cap
Unit Size:
Pack of 12
Pack of 24
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Type unitSize
12084118 250 mL Tamper-evident Screw Cap Pack of 24
12074118 125 mL Tamper-evident Screw Cap Pack of 24
12064118 500 mL Tamper-evident Screw Cap Pack of 12
12094118 1000 mL Tamper-evident Screw Cap Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12084118
Discount Lab Supplies
Use promo code 28170
View all promotions
In Stock
Add to basket
Edge
Add to basket

Reduce the risk of cross contamination with these sterile, disposable media bottles

The Fisherbrand PETG Media Bottle is ideal for media preparation and storage.

  • An inexpensive and shatterproof alternative to glass media bottles
  • Nonpyrogenic and noncytotoxic to protect cells
  • Molded-in graduations won't rub off
  • Made from clear, lightweight, polyethylene terephthalate copolyester (PETG) providing impact resistance and excellent gas-barrier properties
  • Closure is is high density polyethylene (HDPE)
  • Bottles packed in a shrink-wrapped tray to stay clean
  • Heat-shrink band around cap and bottle neck provides visual assurance of sterility for individual bottles until band is removed
  • Part numbers ending in “A” are designed without a temper-evident ring

Compliance

Certified sterile at SAL 10-6, USP Class VI to ensure biocompatibility.

TRUSTED_SUSTAINABILITY

Specifications

Capacity 8.45 oz., 250mL
Capacity (Metric) 250 mL
Capacity (English) 8.45 oz.
Color Clear
For Use With (Application) For media preparation and storage
Material PETG
Sterility Sterile
Certifications/Compliance SAL 10-6, USP Class VI
Closure Included Yes
Disposable Single-use
Graduated Yes
Shape Square
Product Type Media Bottle
Closure Material HDPE
Closure Size 38-430 mm
Closure Type Tamper-evident Screw Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.