missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Autoclavable HD Biohazard Waste Bags

Product Code. 18588056 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Clear with Red Print
Red with Black Print
Dimensions (L x W):
12 x 8 in. (30 x 20 cm)
23 x 19 in. (58 x 48 cm)
24 x 12 in. (61 x 30 cm)
30 x 24 in. (76 x 61 cm)
36 x 24 in. (91 x 61 cm)
Thickness:
1.25 mil
1.50 mil
Unit Size:
Pack of 100
Pack of 1000
Pack of 200
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Dimensions (L x W) Thickness unitSize
18588056 Red with Black Print 12 x 8 in. (30 x 20 cm) 1.25 mil Pack of 1000
18356513 Clear with Red Print 30 x 24 in. (76 x 61 cm) 1.50 mil Pack of 100
18568056 Clear with Red Print 24 x 12 in. (61 x 30 cm) 1.50 mil Pack of 100
18376533 Clear with Red Print 36 x 24 in. (91 x 61 cm) 1.50 mil Pack of 100
18336513 Red with Black Print 23 x 19 in. (58 x 48 cm) 1.25 mil Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 18588056
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Strong and leak-resistant

  • Printed with biohazard symbol and text in English and Spanish
  • Supplied with twist tie closure

Specifications

Closure Type Twist Tie
Color Red with Black Print
Dimensions (L x W) 12 x 8 in. (30 x 20 cm)
Length (Metric) 30 cm
Width (Metric) 20 cm
Autoclavable Autoclavable
Includes Biohazard symbol and text in English and Spanish
Width (English) 8 in.
Length (English) 12 in.
Thickness 1.25 mil
Material Polyethylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.