Learn More
Fisherbrand™ Polypropylene Graduated Specimen Containers
Ideal for specimen collection, transportation and storage
70.30 € - 481.00 €
Product Code | Brand | Capacity (English) | Capacity (Metric) | Closure Included | Sterility | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (English) | Capacity (Metric) | Closure Included | Sterility | Price | Quantity & Availability | ||
11769378
|
Fisherbrand™
|
4.5 oz. | 133 mL | No | Sterile |
234.00 €
Pack of 500 |
Estimated Shipment: 01-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11779378
|
Fisherbrand™
|
5 oz. | 148 mL | Yes | Sterile |
291.00 €
Pack of 100 |
Estimated Shipment: 19-09-2024
Log in to see stock. |
||
11964395
|
Fisherbrand™
|
4.5 oz. | 133 mL | No | Non-sterile |
232.00 €
Pack of 500 |
Estimated Shipment: 11-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11789378
|
Fisherbrand™
|
8 oz. | 237 mL | No | Sterile |
481.00 €
Pack of 500 |
Estimated Shipment: 01-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11799378
|
Fisherbrand™
|
8 oz. | 237 mL | Yes | Sterile |
70.30 €
Pack of 100 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11984375
|
Fisherbrand™
|
8 oz. | 237 mL | No | Non-sterile |
208.00 €
Pack of 500 |
Estimated Shipment: 11-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
- Autoclavable
- Graduated
- With or without nonautoclavable polyethylene snap lid
Order Info
Note: Sterile containers with lids are individually wrapped, and are sterile inside container. Nonsterile and sterile containers without lids are bulk packed.
Specifications
Yes | |
Polypropylene |