missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Reusable Amber Glass Media Bottles

Product Code. 15847001 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity (Metric):
10,000 mL
100 mL
1000 mL
2000 mL
250 mL
500 mL
5000 mL
Unit Size:
Each
Pack of 10
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15847001 500 mL Pack of 10
15817001 100 mL Pack of 10
15837001 250 mL Pack of 10
15827001 1000 mL Pack of 10
17455353 2000 mL Each
17465353 5000 mL Each
17445353 10,000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 15847001
Save Up to 60% on Microbiology Essentials
Use promo code 28004
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use these amber Fisherbrand™ Reusable Glass Media Bottles for a variety of general laboratory applications.

  • Permanent White Enamel Marking Spot
  • GL 45 Screw Thread
  • Linerless
  • Autoclavable (to 140°C)
  • Amber Borosilicate Glass
  • Polypropylene Cap

Specifications

Capacity (Metric) 500 mL
Color Amber
Diameter (English) 3.3 in.
Diameter (English) Inner Neck 1.18 in.
Diameter (Metric) Inner Neck 30 mm
For Use With (Application) Media and reagent storage, Holding and dispensing fluids, Chemical storage
Includes Screw Cap, Pouring Ring
Material Borosilicate Glass (3.3)
No. per Case 10/Cs.
Screw Color Blue
Screw Material Polypropylene
Sterility Non-sterile
Closure Color Blue
Pre-sterilized No
Closure Included Yes
Graduated Yes
Mouth Narrow
Height (Metric) 176 mm
Height (English) 6.9 in.
Closure Material Polypropylene
Closure Size GL 45
Diameter (Metric) 86 mm
Closure Type Screw Cap
Autoclavable Autoclavable
Capacity (English) 17 oz.
Shape Cylindrical
Certifications/Compliance ISO 4796
Disposable Reusable
Product Type Amber Media Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.