missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Sterile Plastic Culture Tubes: Clear Polystyrene

Ideal for tissue culture procedures.

Brand:  Fisherbrand™

Additional Details : Weight : 0.45000kg

 View more versions of this product

Product Code. 16676015

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Ideal for tissue culture procedures

  • Gamma-irradiated and tested for sterility; ready to use
  • No chance of residual contamination
  • All screw-cap tubes have outside threads and unconstricted necks, making filling and pipetting easier
  • All snap caps can be positioned for aerobic or anaerobic applications

Order Info

Sterile caps are available separately; contact Fisher Customer Service.

Specifications

Specifications

Clear
75 mm
Ideal for tissue culture procedures
Sterile
Not Included
5 mL
Polystyrene
Bulk - 1 Pk. of 125
12 mm
Culture Tube
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers