Learn More
Fisherbrand™ SureOne™ Filter Tip Reload Pipette Tips
GREENER_CHOICE
Product Code. p-4252448 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
0.1 to 10 μL
0.1 to 5 μL
10 to 100 μL
2 to 20 μL
20 to 200 μL
Unit Size:
Pack of 960
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11997714 0.1 to 5 μL Pack of 960
11937724 2 to 20 μL Pack of 960
11907724 0.1 to 10 μL Pack of 960
11957724 20 to 200 μL Pack of 960
11947724 10 to 100 μL Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11997714
Add to basket
Edge
Add to basket
Sterile and free of RNase, DNase, ATP, pyrogen, bioburden, PCR inhibitors, endotoxin and DNA
Sterile and free of RNase, DNase, ATP, pyrogen, bioburden, PCR inhibitors, endotoxin and DNA
Packaging Features
  • Offset wells allow filter tips to stack
  • Rigid inserts are multi-channel format ensuring secure tip seal
  • Lot numbered inserts for traceability also include easy to read tip volume and Fisher catalog number
  • Sterile packaging with outer sleeve protects tips from contamination while refilling inserts
  • Environmentally friendly reload packaging reduces plastic waste and saves space
  • Blister sleeve guide rails align tip inserts for smooth and reliable transfer into the rack
Rack Features
  • Flush Fit Inserts sit flush with rack, allowing easy multichannel pipetting
  • Recycle Logo molded into all rack components
  • Notched clasp props lid open for autoclave venting
  • Sure Stack lid rim prevents racks from sliding around when stacked and carted around a lab
  • Safe-Lock latch allows for one-handed open and closing ability and won't pop open when dropped
  • Sure-snap design securely holds insert in place, preventing it from being lifted out while pipetting
  • Flip-lid can be either a hinged or lift-off lid; hinged format does not prevent use of a multichannel pipet
Insert Features
  • Inserts are easy to place onto and remove from rack; clip onto rack for a secure fit
  • Lot traceable—all inserts contain the Fisherbrand part number, tip volume, lot number
  • Indented insert holes inhibit tip movement, stabilizing them for multichannel work
  • Contact Clip helps prevent unnecessary contact with the tips while clipping in inserts
  • Multichannel design; all inserts are 96-well format
  • Robust non-pliable design provides secure seals on every tip
TRUSTED_SUSTAINABILITY
Specifications
Description Micropoint Nano tip; Graduated at 2.5uL; 0.1-5uL
Volume (Metric) 0.1 to 5 μL
Filtered Filtered
Sterility Sterile
Color Clear
Autoclavable Autoclavable
Length (Metric) 31 mm
Format Refill
Tip Style Micro Point, Universal
Disposable Single-use
Graduated Yes
Product Line SureOne
Material Polypropylene
Graduations 2.5 μL
Show More Show Less
As part of our commitment to continuous product improvement, your delivery may contain products with some minor color variation, but their form, fit and function will not be affected. Any questions contact your sales representative.