missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ SureOne™ Micropoint Nano Pipette Tips, Universal Fit
GREENER_CHOICE

Product Code. p-160488 Shop All Fisherbrand Products
Click to view available options
Volume (Metric):
0.1 to 5 μL
Sterility:
Non-sterile
Sterile
Format:
Bulk
Rack
Unit Size:
Pack of 1000
Pack of 960

Product Code. 11987724

Only null left
Add to basket
Edge
Add to basket

For use with micro volume pipetters

 

  • Natural color
  • Precision molded for accuracy in low volume pipetting
  • Certified by independent laboratory – Nonsterile: RNase, DNase, DNA, and Pyrogen Free; Nonsterile: RNase, DNase, DNA, Bioburden, PCR Inhibitor, and pyrogen-free

 

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 0.1 to 5 μL
Sterility Non-sterile
Color Natural
Length (English) 1.25 in.
Length (Metric) 31 mm
Clean Claims DNase, RNase, DNA, Bioburden, and PCR inhibitor Free
Format Bulk
Tip Style Micro Point
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line SureOne
Graduations At 2.5 μL
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.