missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Traceable™ Hi-Accuracy Thermometer

Product Code. 11761445 Shop All Fisherbrand Products
Click to view available options
Includes:
1 Stainless Steel Probe
2 Stainless Steel Probes
Unit Size:
Each

Product Code. 11761445

Only null left
Add to basket
Add to basket

Includes

Probe(s), two AA batteries, Stand, Velcro™ magnetic strips, wall mount, Traceable™ certificate

Monitors inlet and outlet simultaneously. Traceable™ Hi-Accuracy Thermometer records time and date for MIN/MAX readings.

  • Two visual (LEDs) and two audio alarm alerts
  • Alarms programmable in 0.1° increments
  • Probe tip penetration of 0.33 in.
  • Stainless-steel probe with 0.125 in. diameter, 6.25 in. stem length, 9 in. overall length and 5 ft. cable length
  • Available with one probe or two probes
  • Includes 2 AA Batteries, Stand, Velcro™ Magnetic Strips, Wall Mount, Traceable™ Certificate

Certifications

Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.

Compliance

Conforms to ITS-90, International Temperature Standard

Specifications

Certifications/Compliance NIST, ITS-90, ISO 17025, A2LA
Depth (English) 0.75 in.
Depth (Metric) 1.9 cm
Diameter (English) Probe 0.12 in.
Diameter (Metric) Probe 3.05 mm
Dimensions (L x W x H) 9 x 2.75 x 4.25 in. (22.86 x 6.98 x 10.79 cm)
For Use With (Application) Measuring temperatures in water baths, cuvets, gas systems, reagents, incubators, pharmaceuticals, petroleum products, foods, wastewater
Height (English) 4.25 in.
Height (Metric) 10.79 cm
Includes 2 Stainless Steel Probes
Length (English) Cable 5 ft.
Length (English) Probe 6.25 in.
Length (Metric) 230 mm
Length (Metric) Cable 152.4 cm
Length (Metric) Probe 15.87 cm
Material Plastic
Resolution 0.01°C
Weight (English) 5 oz.
Weight (Metric) 141.7 g
Width (English) 2.75 in.
Product Type Traceable Thermometer
Length (English) 9 in.
Max. Temperature (English) 158°F
Max. Temperature (Metric) 70°C
Min. Temperature (English) -58°F
Min. Temperature (Metric) -50°C
Temperature Range (English) -58° to +158°F
Temperature Range (Metric) -50°C to +70°C
Calibrated Yes
Battery Type AA
Accuracy ±0.3°C
Display LED
Width (Metric) 6.98 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.