Learn More
Fisherbrand™ Acid Resistant Plastic Tubs for Ultrasonic Cleaners
Product Code. 10248761 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
Ultrasonic Cleaner, S 300/S 300 H
Ultrasonic Cleaners: S 100, S 100/H; S 120, S 120/H
Ultrasonic Cleaners: S 180, S 180/H
Ultrasonic Cleaners: S 30, S 30/H; S 40, S 40/H; S 60, S 60/H
Unit Size:
Each
Capacity (Metric):
1.3 L
5.8 L
11.0 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize Capacity (Metric)
10248761 Ultrasonic Cleaner, S 300/S 300 H Each -
10177381 Ultrasonic Cleaners: S 100, S 100/H; S 120, S 120/H Each 5.8 L
10320892 Ultrasonic Cleaners: S 30, S 30/H; S 40, S 40/H; S 60, S 60/H Each 1.3 L
10137431 Ultrasonic Cleaners: S 180, S 180/H Each 11.0 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10248761
  • ONLINE EXCLUSIVE PRICE
    252.00 €
    203.21 € / Pack of 1
    Save 48.79 €
    19% Off
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
Provide additional tank protection when using harsh chemical cleaning solutions in the ultrasonic cleaner.
  • Designed to provide additional protection against harsh chemical cleaning solutions
  • Comes with a custom-fit cover to prevent leaks and spills
Specifications
Type Acid Resistant Tub
For Use With (Equipment) Ultrasonic Cleaner, S 300/S 300 H
Includes Plastic cover
Material Plastic