missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ FITC

Product Code. 18769708
Change view
Click to view available options
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
18769708 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18769708 Supplier R&D Systems™ Supplier No. 5440/50
  • ONLINE EXCLUSIVE PRICE
    121.75 €
    109.20 € / 50mg
    Save 12.55 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Green fluorescent dye

FITC is a fluorescent reagent for the labeling of amines. Excitation maximum ∽ 495 nm; emission maximum ∽ 525 nm.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3 ',6 '-Dihydroxy-5-isothiocyanatospiro[isobenzofuran-1(3 H),9 '-[9 H]xanthen]-3-one
CAS 3326-32-7
Synonym Fluorescein-5-Isothiocyanate, FITC Isomer 1
Quantity 50 mg
Product Line Tocris
Target Standard Fluorescein, Coumarin and Rhodamine-based Dyes
Molecular Formula C21H11NO5S
Formula Weight 389.38
Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.