missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress FITC

Product Code. 16467916
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16467916 100 mg 100mg
16421946 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16467916 Supplier MedChemExpress Supplier No. HY66019
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress FITC (Fluorescein 5-isothiocyanate) is a fluorescence probe for the labeling of amines. FITC is a pH- and Cu2+-sensitive fluorescence dye.

Specifications

Quantity 100 mg
Product Type Dye Reagents
Molecular Weight (g/mol) 389.38 g/mol
Solubility DMSO : 50 mg/mL (128.41 mM; Need ultrasonic) ∣ H2O : < 0.1 mg/mL (insoluble)
Content And Storage -20°C, protect from light, stored under nitrogen ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light, stored under nitrogen)
Shipping Condition Room temperature
Target HBV
CAS 3326-32-7
Synonym Fluorescein 5-isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.