missing translation for 'onlineSavingsMsg'
Learn More

SCAT™ Fitting Tool for HPLC Safety Cap

Product Code. 17916970 Shop All SCAT Europe Products
missing translation for 'orderingAttributeHoverText'
Color:
Green
missing translation for 'unitSize'
Each
This item is not returnable. View return policy

Product Code. 17916970

missing translation for 'mfr': SCAT™ 160530

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Connect your HPLC capillaries safely and tightly to your SCAT™ system. SCAT™ PFA fittings are chemically resistant and fit Safety Caps and Safety Waste Caps. Available for different capillary sizes.

SCAT™ fittings are available in the following versions:

  • PFA fitting with integrated ferrule in various colors and diameters
  • Tube connectors in various diameters

Specifications

Type Wrench
Color Green
For Use With (Application) For comfortable connection of the capillaries.
Includes Installation Wrench POM, Hexagonal
For Use With SCAT PFA Fittings
Material POM
Autoclavable Autoclavable
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.