missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ FiveEasy™ Plus FP30 Conductivity Meters

Product Code. 15523380 Shop All Mettler Toledo Products
missing translation for 'orderingAttributeHoverText'
Type:
Conductivy Meter
Meter with Standard Kit
missing translation for 'unitSize'
1 unit
This item is not returnable. View return policy

Product Code. 15523380

missing translation for 'mfr': Mettler Toledo™ 30266944

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

Installation QuickGuide, 1413μS/cm and 12.88mS/cm standard sachets

Compact, benchtop meter provides high quality conductivity measurements with simple click of a button

  • Compact, space saving design with ABS housing
  • Remove sensor holder after use and store on the side of the meter
  • Large, well-structured 4.3 in. segmented LCD display for easy viewing
  • Intuitive button arrangement and simple menu for easy operation
  • RS232 or USB ports to export data directly either to a printer or a PC for further processing
  • One point calibration with three predefined standards, manual cell constant input
  • Mini-DIN sensor input
  • Features analog output, Automatic and manual endpoint recording, Acoustic endpoint signal, Visual endpoint signal
  • Ideal for wide range of laboratory applications
Specifications:
  • Conductivity Range: 0.01μS/cm to 500mS/cm
  • Conductivity Resolution: Automatic range
  • Conductivity Accuracy: ±0.5% of the measured value
  • TDS Range: 0.01mg/L to 300g/L
  • TDS Resolution: Automatic range
  • TDS Accuracy: ±0.5% of the measured value
  • Salinity Range: 0.00 to 42 psu
  • Salinity Resolution: Automatic range
  • Salinity Accuracy: ±0.5% of the measured value
  • Temperature Range: -5° to +105°C (23° to 221°F)
  • Temperature Resolution: 0.1°C
  • Temperature Accuracy: 0.3°C
  • Temperature Compensation: Linear: 0.00%/°C to 10.00%/°C, reference temperature: 20° and 25°C
  • Memory Size: 200 measurements, current calibration
  • Power Supply: 100 to 240V, 50/60Hz, 12 VDC
  • Dimensions: 22.7 x 14.7 x 7cm (9 x 5.7 x 2.7 in.)
  • Weight: 0.63kg (1.4 lb.)
Applications:
  • FP30 Meter only: Suited for applications that require a specific sensor not included in a kit
  • FP30 Standard Kit: Suited for aqueous samples with mid to high conductivity
TRUSTED_SUSTAINABILITY

Specifications

Description FiveEasy Plus FP30-Basic
Includes FP30 Meter, electrode holder, 2 buffer sachets each for pH 1413 mS/cm and 12.88 mS/cm), test certificate
Accuracy (Conductivity) ±0.5% of the measured value
For Use With (Application) Applications that require a specific sensor not included in a kit
Measurement Modes Conductivity
Calibration One point calibration with three predefined standards, manual cell constant input
Range (Conductivity) 0.01 μS/cm to 500 mS/cm
Resolution (Conductivity) Automatic range
Temperature Compensation Linear: 0.00%/°C to 10.00%/°C, reference temperature: 20° and 25°C
Range (Salinity) 0 to 42 psu
Range (TDS) 0 mg/L to 300 mg/L
Temperature Range (Metric) -5°C to +105°C
Temperature Range (English) 32°F to 212°F
Resolution (Temperature) 0.1°C
Accuracy (Temperature) 0.3°C
Display Type LCD
Memory 200 measurements, current calibration
Data Points 200
Data Management Details RS232 or USB ports
Inputs Mini-DIN
Dimensions (L x W x H) 22.7 x 14.7 x 7 cm (9 x 5.7 x 2.7 in.)
Weight (Metric) 0.63 kg
Weight (English) 1.4 lb.
Model FP30
Type Conductivy Meter
Voltage 100/240 V
Frequency 50/60Hz
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.