missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ FiveEasy™ Plus FP30 Conductivity Meters
Beschreibung
Includes
LE703 sensor, installation QuickGuide, 1413μS/cm and 12.88mS/cm standard sachets
- Compact, space saving design with ABS housing
- Remove sensor holder after use and store on the side of the meter
- Large, well-structured 4.3 in. segmented LCD display for easy viewing
- Intuitive button arrangement and simple menu for easy operation
- RS232 or USB ports to export data directly either to a printer or a PC for further processing
- One point calibration with three predefined standards, manual cell constant input
- Mini-DIN sensor input
- Features analog output, Automatic and manual endpoint recording, Acoustic endpoint signal, Visual endpoint signal
- Ideal for wide range of laboratory applications
- Conductivity Range: 0.01μS/cm to 500mS/cm
- Conductivity Resolution: Automatic range
- Conductivity Accuracy: ±0.5% of the measured value
- TDS Range: 0.01mg/L to 300g/L
- TDS Resolution: Automatic range
- TDS Accuracy: ±0.5% of the measured value
- Salinity Range: 0.00 to 42 psu
- Salinity Resolution: Automatic range
- Salinity Accuracy: ±0.5% of the measured value
- Temperature Range: -5° to +105°C (23° to 221°F)
- Temperature Resolution: 0.1°C
- Temperature Accuracy: 0.3°C
- Temperature Compensation: Linear: 0.00%/°C to 10.00%/°C, reference temperature: 20° and 25°C
- Memory Size: 200 measurements, current calibration
- Power Supply: 100 to 240V, 50/60Hz, 12 VDC
- Dimensions: 22.7 x 14.7 x 7cm (9 x 5.7 x 2.7 in.)
- Weight: 0.63kg (1.4 lb.)
- FP30 Meter only: Suited for applications that require a specific sensor not included in a kit
- FP30 Standard Kit: Suited for aqueous samples with mid to high conductivity
Spezifikation
Spezifikation
| Description | FiveEasy Plus FP30-Std |
| Includes | FP30 Meter, LE703 pH sensor, electrode holder, 2 buffer sachets each for pH 1413 mS/cm and 12.88 mS/cm), test certificate |
| Accuracy (Conductivity) | ±0.5% of the measured value |
| For Use With (Application) | Suited for aqueous samples with mid to high conductivity |
| Measurement Modes | Conductivity |
| Calibration | One point calibration with three predefined standards, manual cell constant input |
| Range (Conductivity) | 0.01 μS/cm to 500 mS/cm |
| Resolution (Conductivity) | Automatic range |
| Temperature Compensation | Linear: 0.00%/°C to 10.00%/°C, reference temperature: 20° and 25°C |
| Range (Salinity) | 0 to 42 psu |
| Mehr anzeigen |
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?