missing translation for 'onlineSavingsMsg'
Learn More

Velp Scientifica™ FIWE 3 Fiber Analyzer
BIOPHARMA

Product Code. 15688135 Shop All Velp Products
Change view
Click to view available options
Holds:
3 Samples
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds unitSize
15688135 3 Samples Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15688135 Supplier Velp Scientifica™ Supplier No. SA30520201
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

A fully automatic analyzer for crude and detergent fiber determination according to official reference methods requiring minimum operator time, unique user interface, and cloud connectivity.

  • Perform single or multiple extractions including boiling, rinsing, and filtration steps with maximum reproducibility
  • Suitable for raw fiber extraction, conventionally known as an indigestible residue
  • No sample transfer prevents any possible sample loss
  • Crucibles can also be used as sample vessels during weighing, drying, and washing
  • Simply pre-heat the reagents with RC2 Hot Plate (included), and pour them directly into the system
  • An extremely efficient heating element speeds up analysis, ensuring rapid heating of the sample
  • FIWE Series Analyzers reduce the time required for analysis to two hours, compared to six hours for the manual procedure

Specifications

Holds 3 Samples
Material Epoxy Painted Stainless Steel
Includes FIWE 3, RC 2, Heat Shield, PVC Tube, Glass Reagent Bottle (2), P2 Glass Crucible (3), Crucible Pincer, Crucible Holder, and Inlet Tube
For Use With (Application) Fiber Analysis: Weende Method, Neutral Detergent Fiber (NDF), Acid Detergent Fiber (ADF), and Acid Detergent Lignin (ADL)
Height (Metric) 62 cm
Length (Metric) 39 cm
Width (Metric) 53 cm
Weight (Metric) 35 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.