missing translation for 'onlineSavingsMsg'
Learn More

Velp Scientifica™ FIWE 6 Raw Fiber Analyzer
BIOPHARMA

Product Code. 11775714 Shop All Velp Products
Change view
Click to view available options
Capacity (Metric):
6 x 0.5 to 3 g
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11775714 6 x 0.5 to 3 g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11775714 Supplier Velp Scientifica™ Supplier No. SA30520200
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Uses pre-heated reagents for fiber analysis according to Weende, Van Soest and other official methods. Velp Scientifica™ FIWE 6 Raw Fiber Analyzer measures fiber conentration as crude fiber which still measures pectin.

High Performance

  • Rapid analysis - 2 hours vs. 6 hours with manual systems
  • Reliable results
  • High reproducibility
  • Back-pressure pump prevents clogging of crucibles

Versatile

  • Total raw fiber extraction using Weende technique
  • Neutral and acid detergent fiber with NDF, ADF, Van Soest extractions
  • Acid detergent lignin extraction with ADL and Van Soest methods
  • Fiber fraction analysis (cellulose, hemicellulose and pectin)
  • Links boiling, rinsing and filtration
  • Single or multiple samples can be processed

Full System

  • Reagent preheating on RC 2 hotplate (provided)
  • Same crucibles used for weighing, drying and washing - no loss

Specifications

Capacity (Metric) 6 x 0.5 to 3 g
Material Epoxy Painted Stainless Steel
Includes Digital timer with acoustic signal: 0 to 99minutes
For Use With (Application) Hot and cold extractions
Quantity 1 Ea.
Height (Metric) 620 mm
Length (Metric) 390 mm
Width (Metric) 760 mm
Weight (Metric) 40 kg
Electrical Requirements 230 V 50 Hz 1200 w
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.