missing translation for 'onlineSavingsMsg'
Learn More

Velp Scientifica™ FIWE6 Raw Fiber Extractor
BIOPHARMA

Product Code. 15599341 Shop All Velp Products
Change view
Click to view available options
Height (Metric):
105 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Unit Size
15599341 105 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15599341 Supplier Velp Scientifica™ Supplier No. SA30530200
Estimated Shipment: 23-04-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Velp Scientifica™ FIWE6 Raw Fiber Extractor that uses pre-heated reagents for fiber analysis according to Weende, Van Soest and other official methods.

High Performance

  • Rapid analysis – 2 hours vs. 6 hours with manual systems
  • Reliable results
  • High reproducibility
  • Back-pressure pump prevents clogging of crucibles

Versatile

  • Total raw fiber extraction using Weende technique
  • Neutral & acid detergent fiber with NDF, ADF, Van Soest extractions
  • Acid detergent lignin extraction with ADL & Van Soest methods
  • Fiber fraction analysis (cellulose, hemicellulose & pectin)
  • Links boiling, rinsing and filtration
  • Single or multiple samples can be processed

Full System

  • Reagent preheating on RC 2 hotplate (provided)
  • Same crucibles used for weighing, drying & washing – no loss

Specifications

Material Epoxy Painted Stainless Steel
Includes Heat shield, 6 glass crucible P2, RC2 Hot Plate, Reagent Glass Bottle, Puncerm Crucible holder, Inlet tube
For Use With (Application) Raw fiber analysis
Height (Metric) 105 mm
Width (Metric) 760 mm
Weight (Metric) 40 kg
Electrical Requirements 230 V 50/60 Hz
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.