missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Fixation/Permeabilization Concentrate

Product Code. 12736048
Change view
Click to view available options
Quantity:
30 mL
Unit Size:
30mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12736048 30 mL 30mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Contains formaldehyde
Product Code. 12736048 Supplier Invitrogen™ Supplier No. 00512343
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Contains formaldehyde

Includes

One 30 mL bottle

Intended to be used with antibodies to transcription factors and nuclear proteins, such as Foxp3 and Ki-67, as well as cytokines and chemokines.

  • This must be used in combination with the Fixation/Permeabilization Diluent (cat. 00-5223) and the Permeabilization Buffer (10X) (cat. 00-8333)
  • Alternatively, all buffers required for staining with these antibodies can be purchased as a set in the Foxp3/Transcription Factor Staining Buffer Set (cat. 00-5523)
TRUSTED_SUSTAINABILITY

Specifications

Includes One 30 mL bottle
Quantity 30 mL
Reagent Type Fixation Reagent, Permeabilization Reagent
Product Line eBioscience
Product Type Fixation/Permeabilization Buffer
compliance-icons compliance-icons
Product Identifier
  • eBioscience Fixation/Permeabilization Concentrate
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1
  • Germ cell mutagenicity Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H370-Causes damage to organs.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.