Learn More
Invitrogen™ eBioscience™ Fixation/Permeabilization Diluent
Product Code. 12746048
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
Pack of 100
Product Code. Quantity Unit Size
12746048 100 mL Pack of 100
1 options
Contains Sodium azide
Product Code. 12746048 Supplier Invitrogen™ Supplier No. 00522356
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Contains Sodium azide
Intended to be used with antibodies to transcription factors and nuclear proteins, such as Foxp3 and Ki-67, as well as cytokines and chemokines.
  • This must be used in combination with the Fixation/Permeabilization Concentrate (cat. 00-5123) and the Permeabilization Buffer (10X) (cat. 00-8333)
  • Alternatively, all buffers required for staining with these antibodies can be purchased as a set in the Foxp3/Transcription Factor Buffer Staining Set (cat. 00-5523)
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 mL
Reagent Type Fixation Diluent, Permeabilization Diluent
Product Line eBioscience
Product Type Fixation/Permeabilization Diluent