missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf Fixed-angle Rotor for SpinPro™ 8 R / Centrifuge CR22N

Product Code. 32226680 Shop All Eppendorf Products
Change view
Click to view available options
Capacity:
6 x 1000 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
32226680 6 x 1000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32226680 Supplier Eppendorf Supplier No. 5720411480
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Offers a wide variety of rotors for High-Speed Centrifuges, ensuring the perfect fit for specific applications.

  • Rotor type: fixed-angle rotor
  • Application: For harvesting of bacteria, cells or yeast for e.g. recombinant protein or antibody purification in bottles with 6 x 1 L
  • Max. speed: 6100 x g (5300 rpm)
  • Max. capacity: 6 x 1000 mL (Nominal capacity)
  • Made from high-quality: aluminum/PEI (Polyetherimid)
TRUSTED_SUSTAINABILITY

Specifications

Material Aluminum
Type Fixed Angle Rotor
Capacity 6 x 1000 mL
Max. RCF 6100 x g
Max. Speed 5300 rpm
K Factor Depending On Vessel 7886 or 7357
Biocontainment No
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.