Learn More
Erlab Fixed Base With Castor Wheels - Solis 1800
Product Code. 17786584 Shop All Erlab Products
Change view
Click to view available options
Product Type:
Class II Biosafety Cabinet
Unit Size:
Each
Product Code. Product Type Unit Size
17786584 Class II Biosafety Cabinet Each
1 options
Product Code. 17786584 Supplier Erlab Supplier No. NORT000231
  • ONLINE EXCLUSIVE PRICE
    1380.00 €
    1272.00 € / Pack of 1
    Save 108.00 €
    7% Off
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Adapted base for your Biological Safety Cabinet.
  • With castor wheels
  • Compatible model: BSC Solis 1800
Specifications
Product Type Class II Biosafety Cabinet