missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Transferpettor™ Fixed Volume Pipette

Product Code. 10008540 Shop All Brand Products
Change view
Click to view available options
Volume (Metric):
1 μL
5 μL
10 μL
20 μL
50 μL
100 μL
200 μL
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10008540 200 μL Each
10483921 10 μL Each
10088520 20 μL Each
10232221 5 μL Each
10478302 50 μL Each
10623782 1 μL Each
10509681 100 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 10008540 Supplier BRAND™ Supplier No. 701878
Add to basket
Add to basket
This item is not returnable. View return policy

Ideal for liquids when air displacement pipettes won't work. Brand™ Transferpettor™ Fixed Volume Pipettes handle viscous, foaming liquids and high vapor pressure.

  • High degree of precision and accuracy
  • For the most demanding pipetting operations
  • Operates on the positive displacement principle, with piston in direct contact with the aspirated liquid
  • Tight-fitting piston always wipes the walls of the tips completely clean, providing reproducible results
  • No need to discard tips after each pipetting operation since residual wetting is negligible
  • Suitable for media with density up to 13.6g/cm³, viscosity up to 50,000mm2/s, vapor pressure up to 500 mbar
  • Working temperature range of 15° to 40°C
  • Supplied with performance certificate

TRUSTED_SUSTAINABILITY

Specifications

Accuracy 0.5%, ±1.0μL
Certifications/Compliance DIN EN ISO 8655-2
Color Red
Type Pipette
Volume (Metric) 200 μL
Temperature (Metric) Operating 15°C to 40°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.