Learn More
Thermo Scientific™ Fixed-Needle, Gas-Tight GC Syringe, Pk. Of 6
Product Code. 15284563 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
10 μL
Unit Size:
Each
Product Code. Volume (Metric) Unit Size
15284563 10 μL Each
1 options
Product Code. 15284563 Supplier Thermo Scientific™ Supplier No. 365D0626
Estimated Shipment: 18-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Thermo Scientific™ syringes for GC instruments are designed to provide enhanced durability,clarity and accuracy in sample introduction.
TRUSTED_SUSTAINABILITY
Specifications
Needle Gauge 23 to 26 G
Product Type Gas Tight GC Autosampler Syringe, Fixed Needle
Volume (Metric) 10 μL
Length (Metric) Needle 42 mm
Needle Point Style Cone
For Use With (Equipment) Agilent GC Instruments