Learn More
Cole-Parmer™ Fixed-Position Utility Clamps
Product Code. 11765399 Shop All Cole-Parmer Products
Change view
Click to view available options
Diameter (Metric):
69 mm
77 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
11765399 77 mm Each
11705419 69 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11765399 Supplier Cole-Parmer™ Supplier No. 0802200
Only null left
Add to basket
Add to basket
Used to hold apparatus at a fixed 90 degree angle near the lab frame where no adjustment is required after set-up. Cole-Parmer™ Fixed-Position Utility Clamps are ideal for precise alignment of burettes, thermometers and reflux condensers.
Clamps are supplied with non-slip vinyl sleeves or for temperatures above 100°C, fiberglass covers. Choose single adjustment style for standard use where one prong or finger is adjusted using a wing nut, while the other prong remains stationary or for a more versatile clamp choose the dual adjustment style. Dual adjustment clamps offer greater flexibility over a wider range. Utilizing thumbscrews, prongs are easily adjusted over a wider range. Clamps include an integral connector which accepts rods up to 0.75 in. (19mm) in diameter. An additional holder must be purchased in order to attach clamps to lab-frames, support stands or other apparatus.
Specifications
Quantity 1 Ea.
Type Utility Clamp
Material Nickel-plated Zinc
For Use With (Equipment) Burette
For Use With (Application) Used to hold apparatus at a fixed 90° angle near the lab frame where no adjustment is required after set-up
Diameter (Metric) 77 mm
Length (Metric) 13.3
Size Medium