missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Fixogum Rubber Cement

Product Code. 11430456 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
125 mL
Unit Size:
125mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11430456 125 mL 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11430456 Supplier MP Biomedicals™ Supplier No. 11FIXO0125
Spotlight Offers
Use promo code 28037
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Made for use in the formation of an air tight seal around the perimeter of the glass coverslip during probe hybridization for FISH.

Applications: Fixogum Rubber Cement is made for use in the formation of an air tight seal around the perimeter of the glass coverslip during probe hybridization for FISH.

Specifications

Content And Storage Room Temperature
Format Tube
Product Type Biochemicals
Quantity 125 mL
Form Liquid
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Fixogum Rubber Cement
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Germ cell mutagenicity Category 1A
  • Carcinogenicity Category 1A
  • Aspiration toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H336-May cause drowsiness or dizziness.
  • H304-May be fatal if swallowed and enters airways.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P321-Specific treatment (see on this label).
  • P331-Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Naphtha (petroleum),hydrotreated light

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.