missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies FK506

Product Code. 17190332
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17190332 50 mg 50mg
17180332 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17190332 Supplier STEMCELL Technologies Supplier No. 74154
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Calcineurin/NFAT pathway inhibitor; Binds FKBP-12

  • FK506 is a macrolide antibiotic with potent immunosuppressive properties (Thomson et al.) It inhibits the calcineurin/NFAT pathway (Martinez-Martinez & Redondo) and binds with high affinity to FK506 binding protein 12 (FKBP-12) (Thomson et al.)
  • Promotes the self-renewal of mouse embryonic stem cells independent of leukemia inhibitory factor (LIF), and attenuates differentiation following LIF withdrawal (Li et al.)
  • In combination with 1-Azakenpaullone (Catalog #74252) and ID-8 (Catalog #72502), enhances the expansion rates of several human pluripotent stem cell lines (Yasuda et al.)
  • Inhibits pro-inflammatory cytokine production by activated T cells (Sakuma et al.)

Specifications

CAS 104987-11-3
Synonym Tacrolimus
Quantity 50 mg
Molecular Formula C44H69NO12
Purity ≥99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.