missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAPTA, MedChemExpress
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
Description
BAPTA is a non-permeable, selective extracellular calcium chelator, with 105-fold greater affinity for Ca2+ than Mg2+. BAPTA is a valuable tool to study the role of calcium in cell signaling.
Specifications
Specifications
| Chemical Name or Material | BAPTA |
| CAS | 85233-19-8 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C22H24N2O10 |
| Quantity | 200 mg |
| Solubility Information | 1M NaOH : 100 mg/mL (209.89 mM; ultrasonic and adjust pH to 11 with NaOH) ∣DMSO : 15.62 mg/mL (32.79 mM; ultrasonic and warming and heat to 60°C) |
| SMILES | O=C(O)CN(C1=CC=CC=C1OCCOC2=CC=CC=C2N(CC(O)=O)CC(O)=O)CC(O)=O |
| Molecular Weight (g/mol) | 476.43 |
| Formula Weight | 476.43 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction