Learn More
Macherey-Nagel™ Flash Chromatography Kit I
Product Code. 10515561 Shop All Macherey-Nagel Products
Change view
Click to view available options
Type:
Flash Chromatography Kit I
Unit Size:
Each
Product Code. Type Unit Size
10515561 Flash Chromatography Kit I Each
1 options
Product Code. 10515561 Supplier Macherey-Nagel™ Supplier No. 727450
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Type Flash Chromatography Kit I
Includes Includes One 450 x 20 mm I.D. Glass Column, One 1-L eluent Reservoir, 100 g Silica 60 (40 to 63 μm), Sea Sand, Silanized Glass Fiber Wadding, 1 m PTFE Tubing
For Use With (Application) Low Pressure Flash Chromatography