Learn More
Gas Arc™ Inline Flashback Arrestor
Product Code. 12960820
Encompass_Preferred
Change view
Click to view available options
For Use With (Application):
Fuel Gas
Oxygen
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
12960820 Fuel Gas Each
12950820 Oxygen Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12960820 Supplier Gas Arc™ Supplier No. FBA07048
Estimated Shipment: 18-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Connection Type 3/8 BSP LH Female (inlet); 3/8 BSP LH Male (outlet)
Delivery Pressure 5 bar
Type Inline Flashback Arrestor
For Use With (Application) Fuel Gas
Pressure 5 bar (inlet)/5 bar (outlet)