missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ SILBERBRAND™ Measuring Flask for Sugar Analysis

Product Code. 12943011 Shop All Brand Products
Change view
Click to view available options
Capacity (Metric):
100 mL
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
12943011 100 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12943011 Supplier BRAND™ Supplier No. 402038
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For Kohlrausch sugar analysis. Calibrated to contain (TC, In.)

  • Borosilicate glass (Boro 3.3) construction
  • For use with Kohlrausch sugar analysis
  • Error limits according to Class B
  • Calibrated to contain (TC, In)
TRUSTED_SUSTAINABILITY

Specifications

Calibration To contain (TC, In.)
Capacity (Metric) 100 mL
Material Borosilicate Glass (3.3)
Limit of Error 0.2 mL
No. per Pack 2
Class B
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.