missing translation for 'onlineSavingsMsg'
Learn More

Manutan Flat belt sling, yellow, Duplex, 3000kg capacity, three meter length

Product Code. 12839549
Encompass_Preferred
This item is not returnable. View return policy
Product Code. 12839549 Supplier Manutan Supplier No. A340192

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Safety factor 7:1.

  • Vertical lift SWL : 3000kg
  • Effective working length: 3m
  • Sling colour: yellow
  • Width: 75mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.