missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenz Laborglasintrumente Flat Bottom Flasks, Wide Neck
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 07050037
Additional Details : Weight : 0.01000kg
Description
- Acc. to DIN ISO 24450
- With beaded rim
- Material: DURAN™
Specifications
100 mL | |
Duran Glass | |
105 mm |