Learn More
Lenz Laborglasintrumente Flat Bottom Flasks, Wide Neck
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
146.28 € - 627.52 €
Product Code | Brand | Capacity (Metric) | Height (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Height (Metric) | Price | Quantity & Availability | ||||
10017839
|
Lenz Laborglasintrumente
07050049 |
250 mL | 138 mm |
170.66 €
Pack of 10 |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||||
11913831
|
Lenz Laborglasintrumente
07050037 |
100 mL | 105 mm |
146.28 €
Pack of 10 |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||||
11923831
|
Lenz Laborglasintrumente
07050028 |
50 mL | 90 mm |
350.86 €
Pack of 10 |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||||
11943831
|
Lenz Laborglasintrumente
07050070 |
1 L | 190 mm |
398.56 €
Pack of 10 |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||||
11953831
|
Lenz Laborglasintrumente
07050073 |
2 L | 230 mm |
627.52 €
Pack of 10 |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||||
11963831
|
Lenz Laborglasintrumente
07050074 |
2 L | 240 mm |
627.52 €
Pack of 10 |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||||
Description
- Acc. to DIN ISO 24450
- With beaded rim
- Material: DURAN™
Specifications
Duran Glass |