Learn More
DURAN Flat Flange DN 60
Product Code. 31907797 Shop All DWK Life Sciences Products
Change view
Click to view available options
Includes:
With Groove
Without Groove
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes
31907797 With Groove
31907796 Without Groove
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 31907797 Supplier DURAN Supplier No. 210343479
  • ONLINE EXCLUSIVE PRICE
    96.75 €
    73.45 € / Pack of 2
    Save 23.30 €
    24% Off
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Flat flanges are precision‐ground borosilicate glass components for building pressure‐resistant reaction vessels, delivers reliable sealing and high mechanical strength
  • Manufactured from borosilicate glass 3.3 ground, component for further processing into reaction vessels
  • Due to the geometry and the massive wall thickness, the blanks are suitable for devices with pressure and vacuum applications,glass articles for further processing
  • Available with or without grooves, they deliver reliable sealing, high mechanical strength, and trusted DURAN™ quality for demanding laboratory use.
Specifications
Product Type Flat Flange
For Use With (Equipment) Pressure‐resistant Reaction Vessels
Includes With Groove
Material Borosilicate Glass
Quantity 1 Ea.