Learn More
Lenz Laborglasinstrumente™ Flat Flange Lid with Center Neck, Two Angled Side Necks, and One Vertical Side Neck
Product Code. 11552364
Change view
Click to view available options
Closure Size:
Center Neck (NS): 14/23
Center Neck (NS): 29/32
Center Neck (NS): 45/40
Includes:
Laboratory Flange 100
Laboratory Flange 150
Laboratory Flange 200
Laboratory Flange 60
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Size Includes Unit Size
11552364 Center Neck (NS): 45/40 Laboratory Flange 200 Each
10861841 Center Neck (NS): 29/32 Laboratory Flange 100 Each
10169529 Center Neck (NS): 45/40 Laboratory Flange 100 Each
10053473 Center Neck (NS): 29/32 Laboratory Flange 150 Each
10179529 Center Neck (NS): 45/40 Laboratory Flange 150 Each
11542364 Center Neck (NS): 29/32 Laboratory Flange 200 Each
11532364 Center Neck (NS): 14/23 Laboratory Flange 60 Each
10101984 Center Neck (NS): 29/32 Laboratory Flange 60 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 11552364 Supplier Lenz Laborglasinstrumente™ Supplier No. 06104564
  • ONLINE EXCLUSIVE PRICE
    886.00 €
    629.40 € / Pack of 1
    Save 256.60 €
    28% Off
Estimated Shipment: 18-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Material Borosilicate Glass, Duran Glass
For Use With (Application) Reaction vessels
Includes Laboratory Flange 200
Product Type Flange lid
Closure Size Center Neck (NS): 45/40