missing translation for 'onlineSavingsMsg'
Learn More

Flavin adenine dinucleotide disodium salt, 100.5%, 95 to 99%, MP Biomedicals™

Product Code. 11406942 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11406942 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11406942 Supplier MP Biomedicals Supplier No. 0215112780
  • ONLINE EXCLUSIVE PRICE
    89.70 €
    85.05 € / 100mg
    Save 4.65 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 29-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 146-14-5
Color Orange
Assay Percent Range 95 to 99%
Molecular Formula C27H33N9O15P2
MDL Number MFCD00041835
Quantity 100 mg
Synonym flavin adenine dinucleotide, flavine-adenine dinucleotide, flavin-adenine dinucleotide, fad, flavitan, fademin, flanin f, unii-zc44yti8kk, flamitajin b, adenine-flavin dinucleotide
InChI Key VWWQXMAJTJZDQX-UHFFFAOYNA-N
SMILES CC1=C(C)C=C2N(CC(O)C(O)C(O)COP(O)(=O)OP(O)(=O)OCC3OC(C(O)C3O)N3C=NC4=C(N)N=CN=C34)C3=NC(=O)NC(=O)C3=NC2=C1
IUPAC Name [[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4S)-5-(7,8-dimethyl-2,4-dioxobenzo[g]pteridin-10-yl)-2,3,4-trihydroxypentyl] hydrogen phosphate
Molecular Weight (g/mol) 785.56
PubChem CID 643975
ChEBI CHEBI:16238
Formula Weight 829.6
Percent Purity 100.5%
Physical Form Powder
Chemical Name or Material Flavin adenine dinucleotide disodium salt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.