missing translation for 'onlineSavingsMsg'
Learn More

Flavin adenine dinucleotide disodium salt, MedChemExpress

Product Code. 16483806
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16483806 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16483806 Supplier Medchem Express Supplier No. HYB1654A
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Flavin adenine dinucleotide (FAD) disodium salt is a redox cofactor, more specifically a prosthetic group of a protein, involved in several important enzymatic reactions in metabolism.

Flavin adenine dinucleotide (FAD) disodium salt is a redox cofactor, more specifically a prosthetic group of a protein, involved in several important enzymatic reactions in metabolism.

Specifications

Chemical Name or Material Flavin adenine dinucleotide disodium salt
CAS 84366-81-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C27H31N9Na2O15P2
Quantity 25 mg
Synonym FAD disodium salt FAD-Na2
Solubility Information H2O : 33.33 mg/mL (40.18 mM; Need ultrasonic) ∣DMSO : 5 mg/mL (6.03 mM; ultrasonic and warming and heat to 60°C)
SMILES CC1=CC(N=C2C(N3)=O)=C(C=C1C)N(C[C@@H]([C@@H]([C@@H](COP(OP(OC[C@H]4O[C@@H](N5C=NC6=C(N=CN=C56)N)[C@@H]([C@@H]4O)O)(O[Na])=O)(O[Na])=O)O)O)O)C2=NC3=O
Molecular Weight (g/mol) 829.51
Formula Weight 829.51
Percent Purity 96.8%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.